Learn More
Epredia™ Papanicolaou EA Stains
GSA_VA
Catalog No. 22050212 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
1 gal.
2.5 gal.
Product Type:
EA-36 Papanicolaou Stain
EA-50 Papanicolaou Stain
EA-65 Papanicolaou Stain
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-050-212 1 gal. EA-65 Papanicolaou Stain
22-050-317 1 pt. EA-36 Papanicolaou Stain
22-050-314 1 gal. EA-36 Papanicolaou Stain
22-050-209 1 pt. EA-50 Papanicolaou Stain
22-050-210 1 gal. EA-50 Papanicolaou Stain
22-050-304 2.5 gal. EA-50 Papanicolaou Stain
22-050-211 1 pt. EA-65 Papanicolaou Stain
22-050-303 2.5 gal. EA-65 Papanicolaou Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-050-212 Supplier Epredia™ Supplier No. 75611
Add to Cart
Add to Cart
This item is not returnable. View return policy
Control the color hues and intensities of your work with Epredia™ Papanicolaou Stain EAs, useful for both gynecological and non-gynecological staining.
  • EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
  • EA-65 is used with OG-6 for non-gynecological staining.
  • Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 1 gal.
Product Type EA-65 Papanicolaou Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov