Learn More
Epredia™ Papanicolaou EA Stains
Catalog No. 22050314 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
1 gal.
2.5 gal.
Product Type:
EA-36 Papanicolaou Stain
EA-50 Papanicolaou Stain
EA-65 Papanicolaou Stain
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-050-314 1 gal. EA-36 Papanicolaou Stain
22-050-317 1 pt. EA-36 Papanicolaou Stain
22-050-209 1 pt. EA-50 Papanicolaou Stain
22-050-210 1 gal. EA-50 Papanicolaou Stain
22-050-304 2.5 gal. EA-50 Papanicolaou Stain
22-050-211 1 pt. EA-65 Papanicolaou Stain
22-050-212 1 gal. EA-65 Papanicolaou Stain
22-050-303 2.5 gal. EA-65 Papanicolaou Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-050-314 Supplier Epredia™ Supplier No. 75411
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Control the color hues and intensities of your work with Epredia™ Papanicolaou Stain EAs, useful for both gynecological and non-gynecological staining.
  • EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
  • EA-65 is used with OG-6 for non-gynecological staining.
  • Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
Specifications
Quantity 1 gal.
Product Type EA-36 Papanicolaou Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov