Learn More
Epredia™ Papanicolaou OG-6 and Modified OG Stains
Catalog No. 22050305 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
1 gal.
2.5 gal.
Product Type:
Modified OG Stain
OG-6 Papanicolaou Stain
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-050-305 2.5 gal. OG-6 Papanicolaou Stain
22-050-207 1 pt. OG-6 Papanicolaou Stain
22-050-208 1 gal. OG-6 Papanicolaou Stain
22-050-287 1 gal. Modified OG Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-050-305 Supplier Epredia™ Supplier No. 75225
Add to Cart
Add to Cart
Stain gynecological specimens with Epredia™ Papanicolaou Stain OGs.
  • OG-6 stains the keratinized areas of the cell
  • Modified OG is a Gill modification that minimizes precipitation and reduces staining time
  • Use OG-6 with EA-36 or EA-50 when staining gynecological specimens
  • Use modified OG with EA-65 for staining non-gynecological specimens
Specifications
Quantity 2.5 gal.
Product Type OG-6 Papanicolaou Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov