Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Papanicolaou's Solution 2b Orange II Solution

Catalog No. M1068872500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500mL
2.5L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1068872500 2.5L
M1068870500 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1068872500 Supplier MilliporeSigma™ Supplier No. 1068872500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aids in diagnosis of cytological cancer and cycle

  • Standard method in cancer and hormone cycle diagnosis
  • Smears with keratinized cells under acidic pH conditions
  • Gives intense reddish staining result with mature and keratinized squamous cells
  • Gives gentle yellow-orange staining result with mature and keratinized squamous cells
TRUSTED_SUSTAINABILITY

Specifications

Packaging Plastic Bottle
Quantity 2.5L
Storage Requirements 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.