Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Papanicolaou's Solution 3b Polychromatic

Catalog No. p-7078316 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1L
2.5L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1092721000 1L
M1092722500 2.5L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1092721000 Supplier MilliporeSigma™ Supplier No. 1092721000
Only null left
Add to Cart
Add to Cart

Used for human-medical cell diagnosis

  • Ready to use
  • Staining solution for cytological cancer cycle diagnosis
  • Used for cytological investigation of sample material of human origin
  • Used to demonstrate the differentiation of squamous cells
TRUSTED_SUSTAINABILITY

Specifications

Packaging Plastic Bottle
Packaging Type Plastic Bottle
Quantity 1L
Storage Requirements 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.