Learn More
Papanicolaou Stain Modified EA 65, 250 mL, G-Biosciences
SCI_ED
Catalog No. S001151
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
S001151 250 mL
1 options
Catalog No. S001151 Supplier G-Biosciences Supplier No. 7862252
Add to Cart
Add to Cart
Papanicolaou Stain Modified EA 65 is used for staining of exfoliative cells in human cytologic specimens.
  • It provides coloration to exfoliative cells of vaginal, cervical, prostatic and other body secretions, allowing critical examination of nuclei and cytoplasmic components
Specifications
Quantity 250 mL
Product Type Papanicolaou Stain