Learn More
ENG Scientific Papanicolaou Staining Solutions
Description
- Solutions are made from certified dyes (where applicable)
- Results: nuclei stain blue with clear, sharp detail; cytoplasm stain varying shades of pink, blue, yellow, green, and gray
- Eosin Y, Bismark Brown Y, Light Green SF, Yellowish, Orange G, Ethanol, Phosphotungstic Acid
Specifications
Specifications
| For Use With (Application) | For use on smears of bronchial aspirates, sputum, pleural and peritoneal fluids |
| Includes | Eosin Y, bismark brown Y, light green SF, yellowish, orange G, ethanol, phosphotungstic acid |
| Packaging | Bottle |
| Quantity | 1 x 3800 mL (1 gal.) |
| Product Type | OG-6 Papanicolaou Stain |
For in vitro diagnostic use only.