Learn More
Cayman Chemical Papaverine Analytical Referenc
SDP
Catalog No. 502887626
Encompass_Preferred
An analytical reference standard categorized as a muscle relaxant; has been found as an impurity in seized heroin; intended for research and forensic applications
Catalog No. 50-288-7626 Supplier Cayman Chemical Supplier No. 1001113310 MG
An analytical reference standard categorized as a muscle relaxant; has been found as an impurity in seized heroin; intended for research and forensic applications