Learn More
AdipoGen Papaverine hydrochloride
SDP
Supplier:  AdipoGen CDXP0164G100
Encompass_Preferred
Chemical. CAS 61-25-6. Formula C20H21NO4 . HCl. MW 339.4 . 36.5. Synthetic. Alkaloid antispasmodic drug that most usually treats visceral spasm or vasospasm. It has also been used in the treatment of erectile dysfunction. Smooth muscle relaxant and vasodilator. Phosphodiesterase PDE10A inhibitor. Show to have antiviral and antipsychotic activity.
Catalog No. 50-256-0214