Learn More
AdipoGen Papaverine hydrochloride
SDP
Catalog No. 502562544
Encompass_Preferred
Chemical. CAS 61-25-6. Formula C20H21NO4 . HCl. MW 339.4 . 36.5. Synthetic. Originally isolated from Papaver somniferum. Smooth muscle relaxant and vasodilator. Phosphodiesterase PDE10A inhibitor. Antiviral. Antipsychotic. Potential inducer of browning in WAT.
Catalog No. 50-256-2544 Supplier AdipoGen Supplier No. AGCN20414G005
Chemical. CAS 61-25-6. Formula C20H21NO4 . HCl. MW 339.4 . 36.5. Synthetic. Originally isolated from Papaver somniferum. Smooth muscle relaxant and vasodilator. Phosphodiesterase PDE10A inhibitor. Antiviral. Antipsychotic. Potential inducer of browning in WAT.