Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Papaverine hydrochloride, 99.6%, MP Biomedicals™

Catalog No. ICN19026105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19026105 5 g
ICN19026125 25 g
ICN19026180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19026105 Supplier MP Biomedicals, Inc Supplier No. 0219026105

Chemical Identifiers

CAS 1263-89-4
Molecular Formula C23H47N5O18S
Molecular Weight (g/mol) 713.706
InChI Key LJRDOKAZOAKLDU-JXDXLELXSA-N
Synonym paromomycin sulfate, aminosidine sulfate;paromomycin sulfate salt
PubChem CID 91972180
IUPAC Name (3S,5S,6S)-5-amino-2-(aminomethyl)-6-[(2R,4S,5S)-5-[(2R,5R,6R)-3,5-diamino-2-[(2S,3S,5S)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-hydroxycyclohexyl]oxy-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxyoxane-3,4-diol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)O)N)OC3C(C(C(O3)CO)OC4C(C(C(C(O4)CN)O)O)N)O)O)N.OS(=O)(=O)O

Specifications

Color White
Assay Percent Range ∽ 99%
Quantity 5 g
Formula Weight 375.9
Physical Form Powder
Chemical Name or Material Papaverine Hydrochloride
EINECSNumber 200-502-1
storageNote1 Protect from light
Recommended Storage Store at room temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.