Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Papaverine hydrochloride, 99%

Catalog No. AC207220250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207220250 25 g Glass bottle
AC207220050 5 g Glass bottle
AC207221000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC207220250 Supplier Thermo Scientific Chemicals Supplier No. 207220250
Only null left
Add to Cart

CAS: 61-25-6 | C20H21NO4·ClH | 375.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-25-6
Molecular Formula C20H21NO4·ClH
Molecular Weight (g/mol) 375.85
MDL Number MFCD00012745
InChI Key UOTMYNBWXDUBNX-UHFFFAOYSA-N
Synonym papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan
PubChem CID 6084
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride
SMILES COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl

Specifications

Melting Point 226.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 222
Merck Index 15, 7122
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol and chloroform,practically insoluble in ether
Formula Weight 375.85
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Papaverine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 200-502-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.