Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Papaverine hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014886 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4886 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4886 Supplier MilliporeSigma Supelco Supplier No. PHR1182500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Papaverine hydrochloride is the hydrochloride salt of an opiate derivative called papaverine, a benzylisoquinoline compound. It has been found to relax smooth muscles of blood vessels and is used in the management of occlusivity and spasticity of pulmonary, coronary and peripheral arteries.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY

Specifications

CAS 61-25-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C20H21 NO4 · HCl
MDL Number MFCD00012745
Synonym 6,7-Dimethoxy-1-veratrylisoquinoline hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 375.85
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.