Learn More
Cayman Chemical PapaverInhydrochlorIde 10mg
SDP
Catalog No. 502917055
Encompass_Preferred
An analytical reference standard categorized as a muscle relaxant; has been found as an impurity in seized heroin; intended for research and forensic applications
Catalog No. 50-291-7055 Supplier Cayman Chemical Supplier No. 3936110 MG
An analytical reference standard categorized as a muscle relaxant; has been found as an impurity in seized heroin; intended for research and forensic applications