Learn More
Thomas Scientific Extra roll of paper for B4000-P
SDP
Catalog No. 501458572
Encompass_Preferred
Despite the space-saving exterior dimensions of the Bioclaves, the stainless steel chambers are surprisingly large, accommodating a variety of liquids, media, instruments, glassware, plasticware and other common laboratory items. Both of these benchtop models include a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI. With large digital displays and fully automatic operation, all segments of the cycle (fill, sterilize, exhaust and dry) commence and run to completion by simply pressing the start button. An optional thermal mini-printer is also available for providing a record of sterilization parameters (compatible with BioClave 16 only). BioClave Mini™ Autoclave Oven : The 8 liter BioClave Mini is preprogrammed to meet basic sterilization needs. although the temperature can be selected at either 121°C or 134°C, the sterilization time is fixed. The economical BioClave Mini is the ideal choice wherev
Catalog No. 50-145-8572 Supplier Thomas Scientific Supplier No. 1227W12
Despite the space-saving exterior dimensions of the Bioclaves, the stainless steel chambers are surprisingly large, accommodating a variety of liquids, media, instruments, glassware, plasticware and other common laboratory items. Both of these benchtop models include a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI. With large digital displays and fully automatic operation, all segments of the cycle (fill, sterilize, exhaust and dry) commence and run to completion by simply pressing the start button. An optional thermal mini-printer is also available for providing a record of sterilization parameters (compatible with BioClave 16 only). BioClave Mini™ Autoclave Oven : The 8 liter BioClave Mini is preprogrammed to meet basic sterilization needs. although the temperature can be selected at either 121°C or 134°C, the sterilization time is fixed. The economical BioClave Mini is the ideal choice wherev