Learn More
Essentra Porous Technologies PAPER 8.5X11 CLRM YLW 2500/CS
SDP
Catalog No. NC0475120
Encompass_Preferred
Cleanroom Compatible Copy Machine Paper, Yellow, 8.5 X 11, 250 Sheets/Pack, 10 Packs/Case Essentra Porous Technologies Keeps Critical Environments Clean-Reducing The Risk Of Contamination Before It Can Even Be A Factor. Our Paper Is Specially Designed For Use In Cleanrooms. Each Page Is Impregnated And Coated With A Unique Polymer Formulation That Inhibits Particulate Generation And Chemical Extractables. Control Contamination, Make Cleaning Easier, And Increase Product Yield With Essentra'S Cleanroom Compatible Paper. • Special Anti-Contaminant Polymer Formulation • Dimensionally Stable And Compatible With Wet And Dry Systems
Catalog No. NC0475120 Supplier Essentra Porous Technologies Supplier No. 97177YW8A00
Cleanroom Compatible Copy Machine Paper, Yellow, 8.5 X 11, 250 Sheets/Pack, 10 Packs/Case Essentra Porous Technologies Keeps Critical Environments Clean-Reducing The Risk Of Contamination Before It Can Even Be A Factor. Our Paper Is Specially Designed For Use In Cleanrooms. Each Page Is Impregnated And Coated With A Unique Polymer Formulation That Inhibits Particulate Generation And Chemical Extractables. Control Contamination, Make Cleaning Easier, And Increase Product Yield With Essentra'S Cleanroom Compatible Paper. • Special Anti-Contaminant Polymer Formulation • Dimensionally Stable And Compatible With Wet And Dry Systems