Learn More
Gilson Company Inc 6in Circular Paper Discs, 500/pk
Catalog No. NC1521190
Encompass_Preferred
Circular Paper Discs prevent hot-mix asphalt samples from sticking to baseplates or compaction heads in Marshall or Superpave Gyratory Compaction Molds. The discs are 0.007-0.008in thick and made of 100lb paper stock, and meet ASTM and AASHTO standards. MSA-121 discs are used with 6in and 150mm molds, and sold 500/package.
This item is not returnable. View return policy
Catalog No. NC1521190 Supplier Gilson Company Inc Supplier No. MSA121
Circular Paper Discs prevent hot-mix asphalt samples from sticking to baseplates or compaction heads in Marshall or Superpave Gyratory Compaction Molds. The discs are 0.007-0.008in thick and made of 100lb paper stock, and meet ASTM and AASHTO standards. MSA-121 discs are used with 6in and 150mm molds, and sold 500/package.