Learn More
Med Vet International Exam Table Paper
Catalog No. 502829528
Encompass_Preferred
Exam Table Paper: Medline's strong, absorbent table paper protects exam tables from dirt and moisture while offering comfort and protection for patients. Choose from smooth or crepe finish (crepe is softer, quieter and more absorbent). Crepe is also more textured which will help prevent it from sliding on surfaces. Available in standard and deluxe grades. Deluxe grade features a heavier basis weight paper. All rolls are mechanically metered for exact length, and individually poly bagged for cleanliness during shipment and storage. White. Standard Crepe, 21" x 125 ft.
Catalog No. 50-282-9528 Supplier Med Vet International Supplier No. NON23325H
Exam Table Paper: Medline's strong, absorbent table paper protects exam tables from dirt and moisture while offering comfort and protection for patients. Choose from smooth or crepe finish (crepe is softer, quieter and more absorbent). Crepe is also more textured which will help prevent it from sliding on surfaces. Available in standard and deluxe grades. Deluxe grade features a heavier basis weight paper. All rolls are mechanically metered for exact length, and individually poly bagged for cleanliness during shipment and storage. White. Standard Crepe, 21" x 125 ft.