Learn More
Advantec MFS, Inc Paper Filter 15 cm, 100/pk
Supplier:  Advantec MFS, Inc N05B15.0CM
Encompass_Preferred
Nominal Retention (um): 4. Thickness (mm): 0.21. Flow Time (sec): 195.
Catalog No. NC0930449