Learn More
Industrial Controls PAPER HONYWLL CHART 7DAY MIN=2
Catalog No. NC9616169
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Paper, Recorder; For Honeywell DR4200 recorder; Industrial Controls; Seven-day rotation; Ranges 0/100 and -30/70; 100/Pk
Catalog No. NC9616169 Supplier Industrial Controls Supplier No. 24001661601
This item has a minimum qty of 2 per supplier requirements.
Paper, Recorder; For Honeywell DR4200 recorder; Industrial Controls; Seven-day rotation; Ranges 0/100 and -30/70; 100/Pk