Learn More
GA International Paper Labels for Direct Thermal Printers 0.866" x 0.59" / 22mm x 15mm Rectangle White From +4°C to +70°C (+39°F to +158°F)
Catalog No. 5000571446
Encompass_Preferred
Paper labels for direct thermal printers. Provided in a range of colors, the labels are ideal for use as shipping labels or color coding. For general use and organization in offices, laboratories, warehouses, as well as sorting, logistics, and shipping centers.
Catalog No. 50-005-71446 Supplier GA International Supplier No. DTA140C16WH
Paper labels for direct thermal printers. Provided in a range of colors, the labels are ideal for use as shipping labels or color coding. For general use and organization in offices, laboratories, warehouses, as well as sorting, logistics, and shipping centers.