Learn More
GA International Paper Labels for Thermal-Transfer Printers 0.354" / 9mm Circle Violet From -20°C to +120°C (-4°F to +248°F)
Catalog No. 5000572071
Encompass_Preferred
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.
Catalog No. 50-005-72071 Supplier GA International Supplier No. GPB61NPNOTC12VI
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.