Learn More
GA International Paper Labels for Thermal-Transfer Printers 0.875" x 0.875" / 22.2mm x 22.2mm Rectangle White From -20°C to +120°C (-4°F to +248°F)
Catalog No. 5000571470
Encompass_Preferred
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.
Catalog No. 50-005-71470 Supplier GA International Supplier No. GPB98C35WH
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.