Learn More
GA International Paper Labels for Thermal-Transfer Printers 1" x 0.5" / 25.4mm x 12.7mm Rectangle Orange From -20°C to +120°C (-4°F to +248°F)
Catalog No. 5000572613
Encompass_Preferred
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.
Catalog No. 50-005-72613 Supplier GA International Supplier No. GPB7C13OR
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.