Learn More
GA International Paper Labels for Thermal-Transfer Printers 1" x 0.5" / 25.4mm x 12.7mm Rectangle White From -20°C to +120°C (-4°F to +248°F)
Catalog No. 5000571694
Encompass_Preferred
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.
Catalog No. 50-005-71694 Supplier GA International Supplier No. GPB7C13WH
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.