Learn More
GA International Paper Labels for Thermal-Transfer Printers 2.5" x 0.75" / 63.5mm x 19.1mm Rectangle White From -20°C to +120°C (-4°F to +248°F)
Catalog No. 5000572601
Encompass_Preferred
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.*Not recommended for cryogenic use
Catalog No. 50-005-72601 Supplier GA International Supplier No. GPB68C32WH
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.*Not recommended for cryogenic use