Learn More
GA International Paper Labels for Thermal-Transfer Printers 2" x 2" / 50.8mm x 50.8mm Rectangle White From -20°C to +120°C (-4°F to +248°F)
Catalog No. 5000572226
Encompass_Preferred
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.
Catalog No. 50-005-72226 Supplier GA International Supplier No. GPB27C11WH
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.