Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDC Healthcare™ Paper with Permanent Adhesive

Catalog No. 21403439
Change view
Click to view available options
Dimensions (L x W):
1 x 3 in.
2 x 2 in.
2.35 x 0.87 in.
2.5 x 1.25 in.
3 x 1.62 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
21-403-439 2.5 x 1.25 in.
21-403-435 2.35 x 0.87 in.
21-403-436 1 x 3 in.
21-403-437 2 x 2 in.
21-403-438 3 x 1.62 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21-403-439 Supplier PDC Healthcare™ Supplier No. PM18FG
Add to Cart
Add to Cart

Self-adhesive paper hazard labels for clearly marking 0.1% Formalin solutions

  • Pre-printed with 0.1% FORMALIN text for consistent hazard identification
  • Permanent adhesive ensures labels remain firmly attached to containers
  • Fluorescent pink coloring provides excellent visibility in laboratory settings
  • Paper construction allows for clear printing and durability
  • Roll format with 1,000 labels enables efficient application

Specifications

Adhesive Type Permanent
Coating None
Color Green
Finish Matte
Format Roll
Material Paper
Product Type Label
Sterility Non-sterile
Dimensions (L x W) 2.5 x 1.25 in.
Length (English) 2.5 in
Length (Metric) 6.4 cm
Width (English) 1.25 in.
Width (Metric) 3.2 cm
Weight (Metric) 680.4 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.