Learn More
Electron Microscopy Sciences Absorbent Paper Points, Selection 1
SDP
Catalog No. 50930668
Encompass_Preferred
Machine rolled paper with a tapered end which offers good water absorbency, and assists you in the removal of surplus water from the knife boat, on the block face, and from the grids.
Catalog No. 50-930-668 Supplier Electron Microscopy Sciences Supplier No. 7101101
Machine rolled paper with a tapered end which offers good water absorbency, and assists you in the removal of surplus water from the knife boat, on the block face, and from the grids.