Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Paper Towel Dispenser

Catalog No. 15050316 Shop All Fisherbrand Products
Change view
Click to view available options
Height (English):
8 in.
14.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
15-050-316 14.5 in.
15-050-315 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-050-316
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These Paper Towel Dispensers are constructed from 22 gauge, type 304 stainless steel with a #4 finish

  • These Paper Towel Dispensers are constructed from 22 gauge, type 304 stainless steel with a #4 finish.
  • Shipped with bracket to mount on a pegboard or to the wall.

Specifications

Product Type Towel Dispenser
Material Stainless Steel
Depth (English) 4 in.
Height (English) 14.5 in.
Width (English) 11 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.