Learn More
Electron Microscopy Sciences SmartBoats™ Paper Weigh Boats Small, Brown, 35 x 35 mm
SDP
Catalog No. 501929432
Encompass_Preferred
These weigh boats are made from FSC Certified (FSC Mix) paper (Eco-IN) and will naturally biodegrade when disposed of in the standard waste stream (Eco-OUT). They are much easier to handle than regular weighing papers. The weigh boats control contents the same as a polystyrene weigh boat but the design allows for easier pouring. Compatible with heat applications with a temperature limitation of 210 degree C.
Color: Brown
Dimensions: 35 x 35 mm
Height: 15.8 mm
Catalog No. 50-192-9432 Supplier Electron Microscopy Sciences Supplier No. 7001301
These weigh boats are made from FSC Certified (FSC Mix) paper (Eco-IN) and will naturally biodegrade when disposed of in the standard waste stream (Eco-OUT). They are much easier to handle than regular weighing papers. The weigh boats control contents the same as a polystyrene weigh boat but the design allows for easier pouring. Compatible with heat applications with a temperature limitation of 210 degree C.
Color: Brown
Dimensions: 35 x 35 mm
Height: 15.8 mm