Learn More
Chemglass Life Sciences Paper Weighing Boat, Brown, Medium
SDP
Catalog No. 501532246
Encompass_Preferred
Paper Weighing Boat, Brown, Medium. Eco-Friendly. Will naturally biodegrade. Compatible with heat applications with a temperature limitation of 210C. Medium (4.5 x 4.5 x 1.5cm). Brown for light colored samples.
Catalog No. 50-153-2246 Supplier Chemglass Life Sciences Supplier No. CLS181802
Paper Weighing Boat, Brown, Medium. Eco-Friendly. Will naturally biodegrade. Compatible with heat applications with a temperature limitation of 210C. Medium (4.5 x 4.5 x 1.5cm). Brown for light colored samples.