Learn More
Chemglass Life Sciences Paper Weighing Boat, Brown, Small
SDP
Catalog No. 501532245
Encompass_Preferred
Paper Weighing Boat, Brown, Small. Eco-Friendly. Will naturally biodegrade. Compatible with heat applications with a temperature limitation of 210C. Small (3.5 x 3.5 x 1.5cm). Brown for light colored samples.
Catalog No. 50-153-2245 Supplier Chemglass Life Sciences Supplier No. CLS181801
Paper Weighing Boat, Brown, Small. Eco-Friendly. Will naturally biodegrade. Compatible with heat applications with a temperature limitation of 210C. Small (3.5 x 3.5 x 1.5cm). Brown for light colored samples.