Learn More
Chemglass Life Sciences Paper Weighing Boat, White, Medium
SDP
Catalog No. 501532248
Encompass_Preferred
Paper Weighing Boat, White, Medium. Eco-Friendly. Will naturally biodegrade. Compatible with heat applications with a temperature limitation of 210C. Medium (4.5 x 4.5 x 1.5cm). White for dark samples.
Catalog No. 50-153-2248 Supplier Chemglass Life Sciences Supplier No. CLS181804
Paper Weighing Boat, White, Medium. Eco-Friendly. Will naturally biodegrade. Compatible with heat applications with a temperature limitation of 210C. Medium (4.5 x 4.5 x 1.5cm). White for dark samples.