Learn More
Chemglass Life Sciences Paper Weighing Boat, White, Small
SDP
Catalog No. 501532247
Encompass_Preferred
Paper Weighing Boat, White, Small. Eco-Friendly. Will naturally biodegrade. Compatible with heat applications with a temperature limitation of 210C. Small (3.5 x 3.5 x 1.5cm). White for dark samples.
Catalog No. 50-153-2247 Supplier Chemglass Life Sciences Supplier No. CLS181803
Paper Weighing Boat, White, Small. Eco-Friendly. Will naturally biodegrade. Compatible with heat applications with a temperature limitation of 210C. Small (3.5 x 3.5 x 1.5cm). White for dark samples.