Learn More
Genscript Corporation PAPP-A 1AC2 mAb Mouse
Catalog No. 5000057319
Encompass
Pregnancy-associated plasma protein A (PAPP-A) is a high molecular weight protein htPAPP-A is a protein complex which comprises of two PAPP-A subunits and two proform of eosinophil major basic protein It exists in the blood of pregnant woman dPAPP-A is a htPAPP-A dimmer which is composed of two PAPP-A subunits htPAPP-A serves as a useful marker for the diagnosis of Down syndrome (DS)
Catalog No. 50-000-57319 Supplier Genscript Corporation Supplier No. V01702100
Add to Cart
Add to Cart
Request a Quote
Pregnancy-associated plasma protein A (PAPP-A) is a high molecular weight protein htPAPP-A is a protein complex which comprises of two PAPP-A subunits and two proform of eosinophil major basic protein It exists in the blood of pregnant woman dPAPP-A is a htPAPP-A dimmer which is composed of two PAPP-A subunits htPAPP-A serves as a useful marker for the diagnosis of Down syndrome (DS)