Learn More
Ansh Labs Ansh Labs Recombinant Dimeric PAPP-A (Pregnancy Associated Plasma Protein A) antigen is a HEK-derived antigen and is provided as supernatant. The antigen can be used for binding activity in applications like ELISA or Western blot. Size: 5 ug
SDP
Catalog No. NC3448381
Encompass
We’re currently building out the product information and will be adding additional content soon. Thank you for your patience as we continue to expand our selection and improve your ordering experience.
Catalog No. NC3448381 Supplier Ansh Labs Supplier No. AG301BP024