Learn More
Ansh Labs Ansh Labs PAPP-A ELISA kit, 96-well colorimetric assay. Detection in serum & other fluids in mouse, goat, bovine, canine, equine, feline, ovine, porcine, rat, squirrel monkey & vervet monkey. Range of 0.24-10.3ng/mL & sensitivity of 0.020ng/mL.
SDP
Supplier:  Ansh Labs AL158
Encompass
The Mouse PAPP-A ELISA kit provides materials for the quantitative measurement of PAPP-A (Pregnancy Associated Plasma Protein) in mouse serum, cell supernatant, and other biological fluids, with species reactivity to goat, bovine, canine, canine tissue extract, equine, equine cyst fluid, feline, ovine, porcine, rat, squirrel monkey, and vervet monkey samples. The kit offers an analytical range of 0.24-10.3 ng/mL and a sensitivity of 0.020 ng/mL, using 25 uL of cell culturesupernatant or serum. The kit is an HRP-based ELISA with colorimetric detection by dual wavelength absorbance at 450 nm and 630 nm as reference filter and has an assay time of 3.5 hours. For Research Use Only.
Catalog No. 50-194-3738