Learn More
Ansh Labs PAPP-A2 ELISA 96-wells, 50 L Plasma/Serum, Range 6 calibrators ranging 0.19-16 ng/mL, LOD 0.071 ng/mL, Time 2.5 hours, Shelf Life 24 months
SDP
Supplier:  Ansh Labs AL109
Encompass
PAPP-A2 ELISA has a total 2.5 hours incubation, calibrators and range of 6, 0.19-16 ng/mL, sensitivity of 0.071 ng/mL, sample of 50 µL Serum. Reacts with Human. Crossreactivity with related molecules is not significant.
Catalog No. 50-194-3724