Learn More
MyBioSource PAP ELISA Kit
SDP
Supplier:  MyBioSource MBS250374796STRIPWELLS
Encompass_Preferred
The MyBioSource PAP ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of prostatic acid phosphatase (PAP) in human serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. The kit uses a double-antibody sandwich ELISA to assay PAP levels. It has a detection range of 0.156-10 ng/mL PAP. The sensitivity of this assay is <0.094 ng/mL. The assay takes around 2 hours to complete. This product can detect PAP in multiple sample types. Overall, the MyBioSource PAP ELISA Kit provides a sensitive, quick and convenient way to quantitatively measure PAP concentration in various sample types.
Catalog No. NC2201773