Learn More
Aviva Systems Biology Corp PAPPA2 Antibody - N-terminal region (ARP42555_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42555_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAPPA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PAPPA2. Alternate names: PAPP-A2, PAPPE, PLAC3ApplicationWB. Species reactivity: Guinea Pig, Human, Rat
Catalog No. 50-014-58264