Learn More
Bullard™ PAPR System Breathing Tubes for Loose-Fitting Facepieces
SDP
Catalog No. 19824428 Shop All Bullard Company Products
Change view
Click to view available options
Size:
Regular
X-Long
X-Small
Length (English):
26 in.
38 in.
Length (Metric):
66.04 cm
83.82 cm
96.5 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English) Length (Metric)
19-824-428 X-Long 38 in. 96.5 cm
19-121-906 X-Small 26 in. 66.04 cm
19-021-238 Regular - 83.82 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-824-428 Supplier Bullard™ Supplier No. PA20LFBTXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Breathing tube for use with PA20 and PA30 series powered air-purifying respirator (PAPR) systems.
  • Breathing tube components/replacement parts to build PAPR replace parts for an existing system
  • For use with PA20 and PA30 loose-fitting facepieces
  • Choose from three lengths:
    • Extra small
    • Standard
    • Extra long
Specifications
Product Type Breathing Tube
Size X-Long
For Use With (Equipment) PA20 or PA30 Loose-Fitting Facepieces
Includes Hose clamp
Model PA20LFBTXL
Length (English) 38 in.
Length (Metric) 96.5 cm
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.