Learn More
Cayman Chemical PapRIVtrIfluoROCTAte salt 10mg
SDP
Catalog No. 502918138
Encompass_Preferred
A quorum-sensing peptide; induces IL-6 and TNF-α production by, and NF-κB translocation in, BV-2 microglia cells from 1-25 µM
Catalog No. 50-291-8138 Supplier Cayman Chemical Supplier No. 3745310 MG
A quorum-sensing peptide; induces IL-6 and TNF-α production by, and NF-κB translocation in, BV-2 microglia cells from 1-25 µM