Learn More
AdipoGen Paprotrain (1 mg)
SDP
Supplier:  AdipoGen AGCR13504M001
Encompass_Preferred
Chemical. CAS: 57046-73-8. Formula: C16H11N3. MW: 245.3. Cell permeable and reversible kinesin-specific MKLP-2 (mitotic kinesin-like protein 2) inhibitor. Inhibits the recruitment of the chromosome passenger proteins survivin and aurora B to the central spindle during anaphase chromosome segregation. Moderate DYRK1A inhibitor (IC50= 5.5 µM). Lead compound.
Catalog No. 50-205-3880