Learn More
Medchemexpress LLC HY-101298 5mg Medchemexpress, Paprotrain CAS:57046-73-8 Purity:>98%
SDP
Catalog No. 501932091
Encompass_Preferred
Medchemexpress, HY-101298 5mg Paprotrain CAS:57046-73-8 Paprotrain is a cell-permeable inhibitor of the kinesin MKLP-2 , inhibits the ATPase activity of MKLP-2 with an IC 50 of 1.35 μM and a K i of 3.36 μM and shows a moderate inhibition activity on DYRK1A with an IC 50 of 5.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2091 Supplier Medchemexpress LLC Supplier No. HY1012985MG
Medchemexpress, HY-101298 5mg Paprotrain CAS:57046-73-8 Paprotrain is a cell-permeable inhibitor of the kinesin MKLP-2 , inhibits the ATPase activity of MKLP-2 with an IC 50 of 1.35 μM and a K i of 3.36 μM and shows a moderate inhibition activity on DYRK1A with an IC 50 of 5.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.