Learn More
Abcam Paprotrain, MKLP-2 inhibitor, 1MG
Catalog No. 502705884
Encompass_Preferred
MW 245.28 Da, Purity >98%. Selective cell-permeable MKLP-2 inhibitor. Reversible ATP-noncompetitive ATPase inhibitor (IC₅₀ values are 1.35 and 0.83 μM for basal and microtubule-stimulated activity respectively). Inhibits Aurora B. Induces cell cycle arrest at early metaphase I.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5884 Supplier Abcam Supplier No. AB1443221MG
MW 245.28 Da, Purity >98%. Selective cell-permeable MKLP-2 inhibitor. Reversible ATP-noncompetitive ATPase inhibitor (IC₅₀ values are 1.35 and 0.83 μM for basal and microtubule-stimulated activity respectively). Inhibits Aurora B. Induces cell cycle arrest at early metaphase I.
The product is subject to the following: Abcam Restricted Use Statement