Learn More
TARGETMOL CHEMICALS INC PAQUINIMOD 10MG
SDP
Catalog No. 502576330
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Paquinimod (ABR 25757) is a S100A9 inhibitor preventing S100A9 binding to TLR-4. purity: 99%
Catalog No. 50-257-6330 Supplier TARGETMOL CHEMICALS INC Supplier No. T731010MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Paquinimod (ABR 25757) is a S100A9 inhibitor preventing S100A9 binding to TLR-4. purity: 99%