Learn More
Medchemexpress LLC HY-100442 50mg , Paquinimod CAS:248282-01-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10044250MG
Encompass_Preferred
HY-100442 50mg Paquinimod CAS: 248282-01-1 Paquinimod (ABR 25757) is a specific inhibitor of S100A8/S100A9. Paquinimod rescues the pneumonia with substantial reduction of viral loads in SARS-CoV-2-infected mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9284