Learn More
Medchemexpress LLC Paquinimod-d5 | 248282-01-1 | 98.2% | 355.44 | C21H17D5N2O3 | 1 MG
SDP
Catalog No. 5000214923
Encompass_Preferred
Paquinimod-d5 is a deuterated analog of paquinimod provided as a research-grade labeled standard for analytical and tracer studies. It is intended for use as an internal standard in mass spectrometry and for preclinical investigations of S100A8/S100A9 inhibition. The material is supplied as a high-purity solid suitable for laboratory assays and analytical workflows.
  • Deuterium-labeled analog for tracer and internal standard applications.
  • High purity (98.2%) suitable for analytical assays.
  • Useful in studies targeting S100A8/S100A9 pathways.
  • Appropriate for mass spectrometry and pharmacokinetic assays.
  • Available in small, research-scale quantities as a solid.
Catalog No. 50-002-14923 Supplier Medchemexpress LLC Supplier No. HY100442S11MG
Add to Cart
Add to Cart
Paquinimod-d5 is a deuterated analog of paquinimod provided as a research-grade labeled standard for analytical and tracer studies. It is intended for use as an internal standard in mass spectrometry and for preclinical investigations of S100A8/S100A9 inhibition. The material is supplied as a high-purity solid suitable for laboratory assays and analytical workflows.
  • Deuterium-labeled analog for tracer and internal standard applications.
  • High purity (98.2%) suitable for analytical assays.
  • Useful in studies targeting S100A8/S100A9 pathways.
  • Appropriate for mass spectrometry and pharmacokinetic assays.
  • Available in small, research-scale quantities as a solid.