Learn More
Bachem PAR-1 (1-6) (mouse, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4047522.0005
Encompass_Preferred
PAR-1 (1-6) (mouse, rat) Trifluoroacetate, H-Ser-Phe-Phe-Leu-Arg-Asn-OH, 140436-67-5, C37H54N10O9, Mr 782.90, 5mg. Proteinase Activated Receptor 1 (1-6) (mouse, rat), Thrombin Receptor (1-6) (mouse, rat), SFFLRN. PAR-1 agonist.
Catalog No. 50-260-083